Merck
4,4'-Dihydroxy-trans-stilbene, a resveratrol analogue, exhibited enhanced antioxidant activity and cytotoxicity.
Bioorganic & medicinal chemistry (2009-03-03)
Gui-Juan Fan, Xiao-Da Liu, Yi-Ping Qian, Ya-Jing Shang, Xiu-Zhuang Li, Fang Dai, Jian-Guo Fang, Xiao-Ling Jin, Bo Zhou
ABSTRAKT
Resveratrol (3,5,4'-trans-trihydroxystibene) is a natural phytoalexin present in grapes and red wine, which possesses a variety of biological activities including antioxidant activity. In order to find more active antioxidant with resveratrol as the lead compound we synthesized 4,4'-dihydroxy-trans-stilbene (4,4'-DHS). The antioxidant activities of resveratrol and 4,4'-DHS were evaluated by the reaction kinetics with galvinoxyl radical or Cu(II) ions, and the inhibition effects against free-radical-induced peroxidation of human erythrocyte ghosts. It was found that 4,4'-DHS exhibits remarkably higher antioxidant activity than resveratrol. The oxidative products of resveratrol and 4,4'-DHS in the presence of Cu(II) in acetonitrile were identified as the dihydrofuran dimers by spectroscopic method, and the antioxidant mechanism for 4,4'-DHS was proposed. In addition, 4,4'-DHS exhibits remarkably higher cytotoxicity against human promyelocytic leukemia (HL-60) cells than resveratrol.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)