Merck
  • Mammary epithelial-specific deletion of the focal adhesion kinase gene leads to severe lobulo-alveolar hypoplasia and secretory immaturity of the murine mammary gland.
Mammary epithelial-specific deletion of the focal adhesion kinase gene leads to severe lobulo-alveolar hypoplasia and secretory immaturity of the murine mammary gland.
The Journal of biological chemistry (2007-08-25)
Tamas Nagy, Huijun Wei, Tang-Long Shen, Xu Peng, Chun-Chi Liang, Boyi Gan, Jun-Lin Guan
ABSTRAKT
Integrin-mediated cell adhesion and signaling is required for mammary gland development and functions. As a major mediator of integrin signaling, focal adhesion kinase (FAK) has been implicated to play a role in the survival, proliferation, and differentiation of mammary epithelial cells in previously studies in vitro. To assess the role of FAK in vivo, we created mice in which FAK is selectively deleted in mammary epithelial cells. The mammary gland FAK conditional knock-out (MFCKO) mice are viable, fertile, and macroscopically indistinguishable from the control littermates. In virgin MFCKO mice, mammary ductal elongation is retarded at 5 weeks of age but reaches the full extent by 8 weeks of age compared with the control mice. However, the MFCKO females are unable to nurse their pups due to severe lobulo-alveolar hypoplasia and secretory immaturity during pregnancy and lactation. Analysis of the mammary epithelial cells in MFCKO mice showed reduced Erk phosphorylation, expression of cyclin D1, and a corresponding decrease in proliferative capability compared with the littermate controls. In addition, phosphorylation of STAT5 and expression of whey acidic protein are significantly reduced in the mammary glands of MFCKO mice, suggesting defective secretory maturation in these mice. Therefore, the combination of the severe lobulo-alveolar hypoplasia and defective secretory differentiation is responsible for the inability of the MFCKO females to nurse their pups. Together, these results provide strong support for a role of FAK in the mammary gland development and function in vivo.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.