Merck
The effect of NMDA-R antagonist, MK-801, on neuronal mismatch along the rat auditory thalamocortical pathway.
Scientific reports (2020-07-28)
Gloria G Parras, Catalina Valdés-Baizabal, Lauren Harms, Patricia T Michie, Manuel S Malmierca
ABSTRAKT
Efficient sensory processing requires that the brain maximize its response to unexpected stimuli, while suppressing responsivity to expected events. Mismatch negativity (MMN) is an auditory event-related potential that occurs when a regular pattern is interrupted by an event that violates the expected properties of the pattern. According to the predictive coding framework there are two mechanisms underlying the MMN: repetition suppression and prediction error. MMN has been found to be reduced in individuals with schizophrenia, an effect believed to be underpinned by glutamate N-methyl-D-aspartate receptor (NMDA-R) dysfunction. In the current study, we aimed to test how the NMDA-R antagonist, MK-801 in the anaesthetized rat, affected repetition suppression and prediction error processes along the auditory thalamocortical pathway. We found that low-dose systemic administration of MK-801 differentially affect thalamocortical responses, namely, increasing thalamic repetition suppression and cortical prediction error. Results demonstrate an enhancement of neuronal mismatch, also confirmed by large scale-responses. Furthermore, MK-801 produces faster and stronger dynamics of adaptation along the thalamocortical hierarchy. Clearly more research is required to understand how NMDA-R antagonism and dosage affects processes contributing to MMN. Nonetheless, because a low dose of an NMDA-R antagonist increased neuronal mismatch, the outcome has implications for schizophrenia treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(+)-MK-801 hydrogen maleate, powder