Merck
A Whole Genome-Wide Arrayed CRISPR Screen in Primary Organ Fibroblasts to Identify Regulators of Kidney Fibrosis.
SLAS discovery : advancing life sciences R & D (2020-05-20)
Robert J Turner, Stefan Golz, Carina Wollnik, Nils Burkhardt, Ina Sternberger, Uwe Andag, Hauke Cornils
ABSTRAKT
Kidney fibrosis presents a hallmark of chronic kidney disease. With ever-increasing patient numbers and limited treatment options available, novel strategies for therapeutic intervention in kidney disease are warranted. Fibrosis commonly results from a wound healing response to repeated or chronic tissue damage, irrespective of the underlying etiology, and can occur in virtually any solid organ or tissue. In order to identify targets relevant for kidney fibrosis, we aimed to employ CRISPR screening in primary human kidney fibroblasts. We demonstrate that CRISPR technology can be applied in primary kidney fibroblasts and can furthermore be used to conduct arrayed CRISPR screening using a high-content imaging readout in a whole genome-wide manner. Hits coming out of this screen were validated using orthogonal approaches and present starting points for validation of novel targets relevant to kidney disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Collagen, Type I antibody produced in mouse, clone COL-1, ascites fluid
Sigma-Aldrich
Albumin human, Cellastim S, recombinant, expressed in rice, lyophilized powder, suitable for cell culture, low endotoxin, ≥96% (SDS-PAGE)
Sigma-Aldrich
Saponin from Quillaja sp., Sapogenin content 20-35 %