Merck
Kaempferol Regulates miR-15b/Bcl-2/TLR4 to Alleviate OGD-Induced Injury in H9c2 Cells.
International heart journal (2020-05-19)
Linping Li, Yuanxia Shao, Hongjian Zheng, Heng Niu
ABSTRAKT
Ischemic heart disease (IHD) is one of the world's leading causes of human death. Kaempferol (Kae) was proved to have anti-inflammatory, antioxidant, and anticancer effects. Such properties suggested that it might play protective roles in IHD. In this study, we have attempted to disclose the potential regulating mechanisms of Kae in primary cardiomyocytes and H9c2 cells.Cells were first stimulated by oxygen-glucose deprivation (OGD) and then exposed to Kae. CCK-8 assay and flow cytometry were used to examine cell characteristics. Quantitative reverse-transcription polymerase chain reaction was utilized to test the expression levels of miR-15b and TLR4. Afterward, cell transfection, dual-luciferase activity assay, and western blot were used to explore the potential mechanisms.OGD treatment suppressed cell viability, whereas it enhanced cell apoptosis. Besides, OGD treatment enhanced the expression of apoptosis-associated proteins. Kae exposure, however, attenuated the effects that OGD-induced. Further experiments showed that Kae exposure promoted down-regulation of miR-15b, Bcl-2 and TLR4 were a target of miR-15b. Moreover, Kae enhanced the expression of key factors involved in PI3K/AKT and Wnt/β-catenin pathways, whereas miR-15b mimic reversed the Kae-triggered effects.This investigation revealed that Kae diminished OGD-triggered cell damage through down-regulating miR-15b expression via activating PI3K/AKT and Wnt3a/β-catenin pathways.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-CASP9 (CLEAVED-ASP353), C-Terminal antibody produced in rabbit, affinity isolated antibody