Merck
Robust differentiation of human pluripotent stem cells into endothelial cells via temporal modulation of ETV2 with modified mRNA.
Science advances (2020-08-25)
Kai Wang, Ruei-Zeng Lin, Xuechong Hong, Alex H Ng, Chin Nien Lee, Joseph Neumeyer, Gang Wang, Xi Wang, Minglin Ma, William T Pu, George M Church, Juan M Melero-Martin
ABSTRAKT
Human induced pluripotent stem cell (h-iPSC)-derived endothelial cells (h-iECs) have become a valuable tool in regenerative medicine. However, current differentiation protocols remain inefficient and lack reliability. Here, we describe a method for rapid, consistent, and highly efficient generation of h-iECs. The protocol entails the delivery of modified mRNA encoding the transcription factor ETV2 at the intermediate mesodermal stage of differentiation. This approach reproducibly differentiated 13 diverse h-iPSC lines into h-iECs with exceedingly high efficiency. In contrast, standard differentiation methods that relied on endogenous ETV2 were inefficient and notably inconsistent. Our h-iECs were functionally competent in many respects, including the ability to form perfused vascular networks in vivo. Timely activation of ETV2 was critical, and bypassing the mesodermal stage produced putative h-iECs with reduced expansion potential and inability to form functional vessels. Our protocol has broad applications and could reliably provide an unlimited number of h-iECs for vascular therapies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 40-500 NIH units/mg protein (biuret)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)