Merck
High-resolution transcriptional and morphogenetic profiling of cells from micropatterned human ESC gastruloid cultures.
eLife (2020-11-19)
Kyaw Thu Minn, Yuheng C Fu, Shenghua He, Sabine Dietmann, Steven C George, Mark A Anastasio, Samantha A Morris, Lilianna Solnica-Krezel
ABSTRAKT
During mammalian gastrulation, germ layers arise and are shaped into the body plan while extraembryonic layers sustain the embryo. Human embryonic stem cells, cultured with BMP4 on extracellular matrix micro-discs, reproducibly differentiate into gastruloids, expressing markers of germ layers and extraembryonic cells in radial arrangement. Using single-cell RNA sequencing and cross-species comparisons with mouse, cynomolgus monkey gastrulae, and post-implantation human embryos, we reveal that gastruloids contain cells transcriptionally similar to epiblast, ectoderm, mesoderm, endoderm, primordial germ cells, trophectoderm, and amnion. Upon gastruloid dissociation, single cells reseeded onto micro-discs were motile and aggregated with the same but segregated from distinct cell types. Ectodermal cells segregated from endodermal and extraembryonic but mixed with mesodermal cells. Our work demonstrates that the gastruloid system models primate-specific features of embryogenesis, and that gastruloid cells exhibit evolutionarily conserved sorting behaviors. This work generates a resource for transcriptomes of human extraembryonic and embryonic germ layers differentiated in a stereotyped arrangement.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ROCK Inhibitor (Y-27632), The ROCK Inhibitor (Y-27632) is available in a 5 mg format & has been optimized & validated for cell culture & Neuroscience applications.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)