Merck
BFRs (PBDEs and HBCDs) in freshwater species from Lake Trasimeno (Italy): The singular case of HBCDs in red swamp crayfish.
The Science of the total environment (2020-11-21)
Tamara Tavoloni, Tommaso Stecconi, Roberta Galarini, Simone Bacchiocchi, Ambrosius Josef Martin Dörr, Antonia Concetta Elia, Massimiliano Giannotti, Melania Siracusa, Arianna Stramenga, Arianna Piersanti
ABSTRAKT
Eighty-six samples belonging to five different species (crucian carp, Carassius carassius; European perch, Perca fluviatilis; tench, Tinca tinca; eel, Anguilla anguilla; red swamp crayfish, Procambarus clarkii) collected from Lake Trasimeno (Italy) were analyzed to assess polybrominated diphenyl ethers (PBDEs) and hexabromocyclododecanes (HBCDs) contamination. The Trasimeno is the largest Italian peninsular lake located in Umbria (Central Italy), in a rural area with low anthropogenic impact. All the samples were analyzed by an in-house developed analytical procedure involving a single sample preparation with dual detection: Gas- and Liquid-Chromatography coupled to tandem Mass Spectrometry (GC-MS/MS for PBDEs and LC-MS/MS for HBCDs). BFRs levels in crucian carp, tench and European perch were negligible and mostly below quantification limits (LOQs). In eel, the species with the higher fat content, PBDE sum (15 congeners) ranged from 0.269 to 0.916 ng/g w.w. BDE-47, -100 and -154 accounted for roughly 57%, 16% and 11% of the PBDE sum, respectively, while BDE-99 (usually one of the most abundant congeners in biota), only for 3%. HBCDs (sum of α-, β-, γ-isomers) were found between 0.157 and 1.14 ng/g w.w. with α- as predominant isomer (92% of the sum), followed by γ- (5%) and β- (2%). Peculiar was the contamination in red swamp crayfish characterized by negligible PBDEs and very high HBCDs levels with a singular contamination pattern. In female pools (n = 9) the mean HBCDs sum was 0.150 ng/g w.w., while in males higher concentrations were measured (mean = 2.77 ng/g w.w.). A significant correlation seems to exist between the contamination level and the seasonal cycle only in male crayfish. Interestingly, among the HBCDs, the γ-isomer was the highest (67% of the total) while α- contributes only for 20%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)