Merck
  • Productivity and nutritional and nutraceutical value of strawberry fruits (Fragaria x ananassa Duch.) cultivated under irrigation with treated wastewaters.
Productivity and nutritional and nutraceutical value of strawberry fruits (Fragaria x ananassa Duch.) cultivated under irrigation with treated wastewaters.
Journal of the science of food and agriculture (2020-08-14)
Lapo Renai, Francesca Tozzi, Cristina Va Scordo, Edgardo Giordani, Maria C Bruzzoniti, Donatella Fibbi, Laila Mandi, Naaila Ouazzani, Massimo Del Bubba
ABSTRAKT
Agriculture represents a productive sector typically characterized by a high water demand, whereas freshwater (FW) availability is a problem of increasing concern in the world and FW resources are becoming insufficient for sustaining agricultural irrigation. The reuse of treated wastewaters (TWWs) for crop irrigation could be an efficient tool for reducing water shortage. Hence, the present study evaluated the food quality of Fragaria x ananassa (cultivar Camarosa) fruits irrigated with four types of treated wastewaters (TWWs). Strawberries were analysed for yield, sucrose, fructose, glucose, total soluble polyphenols (TSP), total monomeric anthocyanins (TMA), and antiradical and antioxidant capacity. In addition, a targeted quantification of the most representative phenolic compounds of strawberry was performed. TWWs complied the Italian ministerial decree 185/2003 for wastewater reuse, with very few exceptions, mainly represented by chloride concentrations (258-643 mg L-1 versus a legal threshold of 250 mg L-1 ). The reuse of TWWs reduced fruit yield (10-26%) compared to irrigation with tap water as a control. Irrigation with TWWs gave also rise to the decrease of total sugars (14-26%), TSP (2-10%) and TMA (29-49%). Individual phenolic acids, flavonols and flavanols were quite stable in response to the irrigation with TWWs, whereas anthocyanidins decreased significantly. Although TWWs negatively affected fruit quality, the nutritional and nutraceutical parameters determined in the present study were in line with data previously reported for strawberries purchased in the market or cultivated in research orchards, thus suggesting that the use of TWWs does not prevent the fruit marketability. © 2020 Society of Chemical Industry.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Quercitrin hydrate, ≥78%