Merck
Epstein-Barr virus-derived circular RNA LMP2A induces stemness in EBV-associated gastric cancer.
EMBO reports (2020-08-14)
Li-Ping Gong, Jian-Ning Chen, Min Dong, Zhen-Dong Xiao, Zhi-Ying Feng, Yu-Hang Pan, Yu Zhang, Yu Du, Jing-Yue Zhang, Yuan-Hua Bi, Jun-Ting Huang, Jing Liang, Chun-Kui Shao
ABSTRAKT
Cancer stem cells (CSCs) are cancer-initiating cells that are not only a source of tumorigenesis but also the cause of tumour progression, metastasis and therapy resistance. EBV-associated gastric cancer (EBVaGC) is a distinct subtype of gastric cancer with unique clinicopathological and molecular features. However, whether CSCs exist in EBVaGC, and the tumorigenic mechanism of EBV, remains unclear. Here, NOD/SCID mice were injected subcutaneously with the EBVaGC cell line SNU719 and treated with 5-fluorouracil weekly. Successive generations of xenografts yielded a highly malignant EBVaGC cell line, SNU-4th, which displays properties of CSCs and mainly consists of CD44+ CD24- cells. In SNU-4th cells, an EBV-encoded circRNA, ebv-circLMP2A, expression increased and plays crucial roles in inducing and maintaining stemness phenotypes through targeting miR-3908/TRIM59/p53 axis. Additionally, high expression of ebv-circLMP2A is significantly associated with metastasis and poor prognosis in patients with EBVaGC. These findings not only provide evidence for the existence of CSCs in EBVaGC and elucidate the pathogenic mechanism of ebv-circLMP2A in EBVaGC, but also provide a promising therapeutic target for EBVaGC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Sodium acetate buffer solution, pH 5.2±0.1 (25 °C), Molecular Biology, 3 M, 0.2 μm filtered
Sigma-Aldrich
Phenol – chloroform – isoamyl alcohol mixture, BioUltra, Molecular Biology, 25:24:1