Merck
Fragment-based deconstruction of Bcl-xL inhibitors.
Journal of medicinal chemistry (2010-03-03)
Sarah Barelier, Julien Pons, Olivier Marcillat, Jean-Marc Lancelin, Isabelle Krimm
ABSTRAKT
Fragment-based drug design consists of screening low-molecular-weight compounds in order to identify low-affinity ligands that are then modified or linked to yield potent inhibitors. The method thus attempts to build bioactive molecules in a modular way and relies on the hypothesis that the fragment binding mode will be conserved upon elaboration of the active molecule. If the inverse process is considered, do the fragments resulting from the deconstruction of high-affinity inhibitors recapitulate their binding mode in the large molecule? Few studies deal with this issue. Here, we report the analysis of 22 fragments resulting from the dissection of 9 inhibitors of the antiapoptotic protein Bcl-x(L). To determine if the fragments retained affinity toward the protein and identify their binding site, ligand-observed and protein-observed NMR experiments were used. The analysis of the fragments behavior illustrates the complexity of low-affinity protein-ligand interactions involved in the fragment-based construction of bioactive molecules.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Phenylphenol, 99%
Supelco
2-Phenylphenol, PESTANAL®, analytical standard