Merck
  • A class of 3S-2-aminoacyltetrahydro-beta-carboline-3-carboxylic acids: their facile synthesis, inhibition for platelet activation, and high in vivo anti-thrombotic potency.
A class of 3S-2-aminoacyltetrahydro-beta-carboline-3-carboxylic acids: their facile synthesis, inhibition for platelet activation, and high in vivo anti-thrombotic potency.
Journal of medicinal chemistry (2010-03-25)
Jiawang Liu, Xueyun Jiang, Ming Zhao, Xiaoyi Zhang, Meiqing Zheng, Li Peng, Shiqi Peng
ABSTRAKT
3S-Tetrahydro-beta-carboline-3-carboxylic acid (TCCA) effectively inhibits ADP-induced platelet activation. This paper used TCCA as a lead, modified its 2-position with amino acids, and provided 20 novel 3S-2-aminoacyl-1,2,3,4-tetrahydro-beta-carboline-3-carboxylic acids (5a-t). With the in vitro assay, it was demonstrated that this modification diminished the IC(50) values from 701 nM of TCCA to 10 nM of 5a-t. With the in vivo assay, it was demonstrated that this modification reduced the efficacious dose from 5.0 micromol/kg of TCCA to 0.1 micromol/kg of 5a-t. Comparing the Cerius based conformation of them with that of their analogues, the 3-position modified TCCA, it was suggested that the comparatively unfolded conformation was one of the important factors of enhancing the in vivo antithrombotic potency.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Norharmane, crystalline
Sigma-Aldrich
Harmane, 98%
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Harmine, 98%