Merck
Microfluidic Chip-Based Induced Phase Separation Extraction as a Fast and Efficient Miniaturized Sample Preparation Method.
Molecules (Basel, Switzerland) (2020-12-31)
Yao Shen, Bo Chen, Han Zuilhof, Teris A van Beek
ABSTRAKT
Induced phase separation extraction (IPSE) is an efficient sample clean-up technique that can replace liquid-liquid extraction (LLE). The purpose of this study was to miniaturize IPSE by carrying it out in a microfluidic chip. An IPSE chip was designed and evaluated for its ability to separate and purify samples on a microscale. The 5 × 2 cm chip was fed with a solution of polar to non-polar model compounds in acetonitrile-water (1:1). In the 100 µm wide and 40 µm deep microchannels, the sample solution was efficiently separated into two immiscible phases by adding a hydrophobic solvent as inducer. Analytes present in the sample solution each migrated to their own favorable phase upon phase separation. After optimization, extraction and fractionation were easily and efficiently achieved. The behavior of analytes with a pH-dependent partitioning could be influenced by adjusting the pH of the sample solution. Scutellaria baicalensis extract, used in Traditional Chinese Medicine (TCM), was successfully separated in aglycones and glycosides. In this microscale system, the sample and solvent consumption is reduced to microliters, while the time needed for the sample pretreatment is less than one minute. Additionally, the extraction efficiency can reach up to 98.8%, and emulsion formation is avoided.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Alizarin, Dye content 97 %
Rutin, phyproof® Reference Substance