- A certain proportion of docosahexaenoic acid tends to revert structural and dynamical effects of cholesterol on lipid membranes.
A certain proportion of docosahexaenoic acid tends to revert structural and dynamical effects of cholesterol on lipid membranes.
Biochimica et biophysica acta. Biomembranes (2021-02-12)
V I Pedroni, M B Sierra, L M Alarcón, A R Verde, G A Appignanesi, M A Morini
PMID33571481
ABSTRAKT
This work investigates how docosahexaenoic acid (DHA) modifies the effect of Cholesterol (Chol) on the structural and dynamical properties of dipalmitoylphosphatidylcholine (DPPC) membrane. We employ low-cost and non-invasive methods: zeta potential (ZP), conductivity, density, and ultrasound velocity, complemented by molecular dynamics simulations. Our studies reveal that 30% of DHA added to the DPPC-Chol system tends to revert Chol action on a model lipid bilayer. Results obtained in this work shed light on the effect of polyunsaturated fatty acids - particularly DHA - on lipid membranes, with potential preventive applications in many diseases, e.g. neuronal as, Alzheimer's disease, and viral, as Covid-19.