Merck
  • Isoform-selective inhibition of chrysin towards human cytochrome P450 1A2. Kinetics analysis, molecular docking, and molecular dynamics simulations.
Isoform-selective inhibition of chrysin towards human cytochrome P450 1A2. Kinetics analysis, molecular docking, and molecular dynamics simulations.
Bioorganic & medicinal chemistry letters (2010-09-14)
Lin He, Fan He, Huichang Bi, Jiankang Li, Su Zeng, Hai-Bin Luo, Min Huang
ABSTRAKT
Our kinetics studies demonstrated that the nature product chrysin exhibited a high inhibitory affinity of 54 nM towards human cytochrome P450 1A2 and was comparable to α-naphthoflavone (49 nM), whereas it represented a moderate affinity of 5225 nM against human cytochrome P450 2C9. However, it remains unclear how this inhibitor selectively binds 1A2. To better understand the isoform selectivity of chrysin, molecular docking and molecular dynamics simulations were performed. Chrysin formed a strong H-bond with Asp313 of 1A2. The stacking interactions with Phe226 also contributed to its tight binding to 1A2. The larger and much more open active site architectures of 2C9 may explain the weaker inhibitory affinity of chrysin towards 2C9. The predicted binding free energies suggest that chrysin preferred 1A2 (ΔG(bind, pred)=-23.11 kcal/mol) to 2C9 (-20.41 kcal/mol). Additionally, the present work revealed that 7-hydroxy-flavone bound to 1A2 in a similar pattern as chrysin and represented a slightly less negative predicted binding free energy, which was further validated by our kinetics analysis (IC(50)=240 nM). Results of the study can provide insight for designing novel isoform-selective 1A2 inhibitors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Naphthoflavone, ≥98%
Sigma-Aldrich
Chrysin, ≥96.5%