Merck
  • Proteasome inhibition in human breast cancer cells with high catechol-O-methyltransferase activity by green tea polyphenol EGCG analogs.
Proteasome inhibition in human breast cancer cells with high catechol-O-methyltransferase activity by green tea polyphenol EGCG analogs.
Bioorganic & medicinal chemistry (2010-01-05)
Congde Huo, Huanjie Yang, Qiuzhi Cindy Cui, Q Ping Dou, Tak Hang Chan
ABSTRAKT
A pro-drug 8 of a synthetic analog 7 is more active in its antiproliferative activity against human breast cancer MDA-MB-231 cells possessing high catechol-O-methyltransferase (COMT) activity than the pro-drugs of EGCG and the analog 5. The higher activity of 8 is attributed to it not being a substrate of COMT.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
(−)-Epigallocatechin, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%