Merck
  • Searching for multi-target antipsychotics: Discovery of orally active heterocyclic N-phenylpiperazine ligands of D2-like and 5-HT1A receptors.
Searching for multi-target antipsychotics: Discovery of orally active heterocyclic N-phenylpiperazine ligands of D2-like and 5-HT1A receptors.
Bioorganic & medicinal chemistry (2010-02-16)
Gilda Neves, Ricardo Menegatti, Camila B Antonio, Luiza R Grazziottin, Renan O Vieira, Stela M K Rates, François Noël, Eliezer J Barreiro, Carlos A M Fraga
ABSTRAKT
We described herein the design, synthesis, and pharmacological evaluation of N-phenylpiperazine heterocyclic derivatives as multi-target compounds potentially useful for the treatment of schizophrenia. The isosteric replacement of the heterocyclic ring at the biaryl motif generating pyrazole, 1,2,3-triazole, and 2-methylimidazole[1,2-a]pyridine derivatives resulted in 21 analogues with different substitutions at the para-biaryl and para-phenylpiperazine positions. Among the compounds prepared, 4 (LASSBio-579) and 10 (LASSBio-664) exhibited an adequate binding profile and a potential for schizophrenia positive symptoms treatment without cataleptogenic effects. Structural features of this molecular scaffold are discussed regarding binding affinity and selectivity for D(2)-like, 5-HT(1A), and 5-HT(2A) receptors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Phenylpiperazine, ≥97.0%