Merck
Shenmai Injection Upregulates Heme Oxygenase-1 to Confer Protection Against Severe Acute Pancreatitis.
The Journal of surgical research (2020-07-28)
Fei-Hu Zhang, Yang Liu, Xiao-Bin Dong, Hao Hao, Kai-Liang Fan, Xian-Qing Meng, Li Kong
ABSTRAKT
To explore the mechanism of Shenmai injection (SMI) on severe acute pancreatitis (SAP) through heme oxygenase-1 (HO-1) signaling. A total of 40 male Sprague-Dawley (SD) rats (220-260 g) were grouped into the following four categories (n = 10): SAP + SMI + Zinc protoporphyrin (ZnPP), SAP + SMI, SAP, and sham surgery groups. ZnPP is a specific inhibitor of HO-1. Four percent of sodium taurocholate (1 mL/kg) was retrogradely injected via the pancreatic duct to induce the SAP model. The SAP group rats received 1.6 mL/kg saline by intravenous injection 30 min after the induction of SAP. The SAP + SMI group rats received 1.6 mL/kg SMI by intravenous injection 30 min after the induction of SAP. The SAP + SMI + ZnPP group rats received an intravenous injection of 1.6 mL/kg SMI and intraperitoneal administration of 30 mg/kg ZnPP 30 min after the SAP induction. Twenty-four hours after the SAP induction, blood samples were collected for the measurement of amylase, lipase, creatinine, myeloperoxidase, interleukin-10 (IL-10), tumor necrosis factor-α (TNF-α), and HO-1 level, while tissue specimens were harvested for the determination of HO-1, TNF-α, and IL-10 mRNA level. Meanwhile, histopathological changes in organs (pancreas, lung, and kidney) were stored. The serum concentration of amylase, lipase, creatinine, and myeloperoxidase was higher in the SAP group than in the SAP + SMI group. Treatment with SMI increased HO-1 and IL-10 level and reduced TNF-α level in serum and tissues compared to the SAP group (P < 0.05). Treatment with SMI abolished the organ-damaging effects of SAP (P < 0.05). Furthermore, suppression of HO-1 expression by ZnPP canceled the aforementioned effects. SMI confers protection against the SAP-induced systemic inflammatory response and multiple organs damage via HO-1 upregulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard