Merck
Synthesis, molecular modeling and biological evaluation of dithiocarbamates as novel antitubulin agents.
Bioorganic & medicinal chemistry (2010-05-25)
Yong Qian, Gao-Yuan Ma, Ying Yang, Kui Cheng, Qing-Zhong Zheng, Wen-Jun Mao, Lei Shi, Jing Zhao, Hai-Liang Zhu
ABSTRAKT
A series of novel dithiocarbamate compounds with the chalcone scaffold have been designed and synthesized, and their biological activities were also evaluated as potential antiproliferation and antitubulin polymerization inhibitors. Compound 2n showed the most potent biological activity in vitro, which inhibited the growth of MCF-7 cells with IC(50) of 0.04+/-0.01 microM and the polymerization of tubulin with IC(50) of 6.8+/-0.6 microM. To understand the tubulin-inhibitor interaction and the selectivity of the most active compound towards tubulin, molecular modeling studies were performed to dock compound 2n into the colchicine binding site, which suggested probable inhibition mechanism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Colchicine, suitable for plant cell culture, BioReagent, ≥95% (HPLC)