Merck
  • Novel γ-cyclodextrin-metal-organic frameworks for encapsulation of curcumin with improved loading capacity, physicochemical stability and controlled release properties.
Novel γ-cyclodextrin-metal-organic frameworks for encapsulation of curcumin with improved loading capacity, physicochemical stability and controlled release properties.
Food chemistry (2021-01-15)
Yulu Chen, Kedong Tai, Peihua Ma, Jiaqi Su, Wenxia Dong, Yanxiang Gao, Like Mao, Jinfang Liu, Fang Yuan
ABSTRAKT
A safe and biodegradable γ-cyclodextrin-metal-organic-frameworks (γ-CD-MOFs) was successfully synthesized by using an improved hydrothermal method. In this study, curcumin (Cur) was chosen for testing the encapsulation stability and release performance of γ-CD-MOFs. Results of the crystal structure measurement indicated that the encapsulated curcumin within γ-CD-MOFs via van der Waals forces, hydrophobic interactions and hydrogen bonding was failed to disturb the inherent microtopography and crystallinity of γ-CD-MOFs. Compared to individual γ-CD, the γ-CD-MOFs exhibited improved loading capacity, physicochemical stability as well as controlled-release property in simulated digestion, and hence can be regarded as effective carriers for curcumin. Curcumin-loaded γ-CD-MOFs with a Cur : γ-CD-MOFs mass ratio of 2:3 (Cur-CD-MOFs/3), which showed the highest encapsulation efficiency (67.31 ± 2.25%), improved physicochemical stability and controlled-release performance, was selected for further research and industrialization. Our results demonstrate that γ-CD-MOFs can be regarded as a promising novel carrier for the delivery of curcumin or other hydrophobic nutraceuticals.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications