Merck
Structure and biological evaluation of novel cytotoxic sterol glycosides from the marine red alga Peyssonnelia sp.
Bioorganic & medicinal chemistry (2010-11-03)
An-Shen Lin, Sebastian Engel, Benjamin A Smith, Craig R Fairchild, William Aalbersberg, Mark E Hay, Julia Kubanek
ABSTRAKT
Bioactivity-guided fractionation of the extract from a Fijian red alga Peyssonnelia sp. led to the isolation of two novel sterol glycosides 19-O-β-d-glucopyranosyl-19-hydroxy-cholest-4-en-3-one (1) and 19-O-β-d-N-acetyl-2-aminoglucopyranosyl-19-hydroxy-cholest-4-en-3-one (2), and two known alkaloids indole-3-carboxaldehyde (3) and 3-(hydroxyacetyl)indole (4). Their structures were characterized by 1D and 2D NMR and mass spectral analysis. The sterol glycosides inhibited cancer cell growth with mean IC₅₀ values (for 11 human cancer cell lines) of 1.63 and 1.41μM for 1 and 2, respectively. The most sensitive cancer cell lines were MDA-MB-468 (breast) and A549 (lung), with IC₅₀'s in of 0.71-0.97μM for 1 and 2. Modification of the sterol glycoside structures revealed that the α,β-unsaturated ketone at C-3 and oxygenation at C-19 of 1 and 2 are crucial for anticancer activity, whereas the glucosidic group was not essential but contributed to enhanced activity against the most sensitive cell lines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(+)-4-Cholesten-3-one, 98%