Merck
Transcriptional Repression of SIRT3 Potentiates Mitochondrial Aconitase Activation to Drive Aggressive Prostate Cancer to the Bone.
Cancer research (2020-10-30)
Abhisha Sawant Dessai, Mayrel Palestino Dominguez, Uan-I Chen, John Hasper, Christian Prechtl, Cuijuan Yu, Eriko Katsuta, Tao Dai, Bokai Zhu, Sung Yun Jung, Nagireddy Putluri, Kazuaki Takabe, Xiang H-F Zhang, Bert W O'Malley, Subhamoy Dasgupta
ABSTRAKT
Metabolic dysregulation is a known hallmark of cancer progression, yet the oncogenic signals that promote metabolic adaptations to drive metastatic cancer remain unclear. Here, we show that transcriptional repression of mitochondrial deacetylase sirtuin 3 (SIRT3) by androgen receptor (AR) and its coregulator steroid receptor coactivator-2 (SRC-2) enhances mitochondrial aconitase (ACO2) activity to favor aggressive prostate cancer. ACO2 promoted mitochondrial citrate synthesis to facilitate de novo lipogenesis, and genetic ablation of ACO2 reduced total lipid content and severely repressed in vivo prostate cancer progression. A single acetylation mark lysine258 on ACO2 functioned as a regulatory motif, and the acetylation-deficient Lys258Arg mutant was enzymatically inactive and failed to rescue growth of ACO2-deficient cells. Acetylation of ACO2 was reversibly regulated by SIRT3, which was predominantly repressed in many tumors including prostate cancer. Mechanistically, SRC-2-bound AR formed a repressive complex by recruiting histone deacetylase 2 to the SIRT3 promoter, and depletion of SRC-2 enhanced SIRT3 expression and simultaneously reduced acetylated ACO2. In human prostate tumors, ACO2 activity was significantly elevated, and increased expression of SRC-2 with concomitant reduction of SIRT3 was found to be a genetic hallmark enriched in prostate cancer metastatic lesions. In a mouse model of spontaneous bone metastasis, suppression of SRC-2 reactivated SIRT3 expression and was sufficient to abolish prostate cancer colonization in the bone microenvironment, implying this nuclear-mitochondrial regulatory axis is a determining factor for metastatic competence. SIGNIFICANCE: This study highlights the importance of mitochondrial aconitase activity in the development of advanced metastatic prostate cancer and suggests that blocking SRC-2 to enhance SIRT3 expression may be therapeutically valuable. GRAPHICAL ABSTRACT: http://cancerres.aacrjournals.org/content/canres/81/1/50/F1.large.jpg.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Mitochondria Isolation Kit, sufficient for 50 applications (2-5 x 107 cells), isolation of enriched mitochondrial fraction from cells
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)