Merck
Synthesis of novel superdisintegrants for pharmaceutical tableting based on functionalized nanocellulose hydrogels.
International journal of biological macromolecules (2020-11-30)
Shabnam Sheikhy, Ali Akbar Safekordi, Marjan Ghorbani, Khosro Adibkia, Hamed Hamishehkar
ABSTRAKT
Superdisintegrants have an important function in Fast dissolving tablets (FDT). It's believed that an increase in surface to the mass (size reduction) can enhance their performance. Due to the obligation of pharmaceutical excipients being in GRAS (generally recognized as safe) list, we've devoted our research to modify one of the routinely used and important natural polymer, cellulose, as superdisintegrant. Nanocrystalline cellulose (NCC) was extracted from microcrystalline cellulose (MCC) via the sulfuric acid hydrolysis process. NCC derivatives have been synthesized by Itaconic acid/Hydroxyethyl methacrylate (IA/HEMA) via maleic anhydride (MA) to acquire unique swellability properties in to achieve superabsorbent cellulose-based nano hydrogel with the cross-linking system. The disintegration performance of prepared tablets was compared with tablets composed of sodium starch glycolate (SSG) and MCC as positive and negative controls. The results show that the disintegration time of tablets formulated with synthesized modified NCC (m-NCC) decreased dramatically compared to other disintegrants. The dissolution analysis showed suitable condition for complete drug release in a shorter time. The in vitro cytotoxic experiments proved the biocompatibility of newly synthesized superdisintegrant. The dissolution Analysis findings suggest that our developed novel superdisintegrant paves the way for the formulation of fast dissolving tablets containing rapidly acting medicines such as zolpidem.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,2,3,4-Butanetetracarboxylic acid, 99%