Merck
FastFung: A novel medium for the culture and isolation of fastidious fungal species from clinical samples.
Journal of microbiological methods (2020-11-25)
Fadi Bittar, Frédérique Gouriet, Saber Khelaifia, Didier Raoult, Stéphane Ranque
ABSTRAKT
We developed a novel culture medium, referred to FastFung medium as suitable for the culture of clinical fungi, including fastidious ones, for both research and diagnostic studies. It is based on Schædler agar supplemented with many essential components for the growth of fastidious fungi. It also contains selective antibacterial agents for the inhibition of contaminant bacteria growth. In this preliminary study, the FastFung medium was compared to the gold standard Sabouraud medium for 98 fungal and 20 bacterial strains. The fungal strain positive culture rate was 100% vs. 95% and the bacterial strain inhibition was 100% vs. 20%, for the FastFung and Sabouraud media, respectively. When compared to the Sabouraud medium on 120 clinical samples, the FastFung medium displayed both a higher fungal colonies count, and a lower culture contamination rate. Storage at 4 °C for 4 weeks did not alter the FastFung culture medium performances for the six isolates of Candida, Cryptococcus, and Penicillium tested. These encouraging results suggest future development of using the FastFung medium in clinical mycology and in mycobiome characterization. Further prospective evaluation aiming at assessing whether implementing the FastFung medium in the routine workflow simplifies and strengthen fungal isolation capacities in the clinical laboratory is warranted.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Imipenem, Pharmaceutical Secondary Standard; Certified Reference Material