Merck
Untargeted SPME-GC-MS Characterization of VOCs Released from Spray Paint.
Journal of chromatographic science (2020-11-05)
Mitchell Tiessen, Naomi L Stock, Theresa Stotesbury
ABSTRAKT
Paints are a common form of physical evidence encountered at crime scenes. This research presents an optimized method for the untargeted analysis of volatile organic compounds (VOCs) in spray paint using solid-phase microextraction-gas chromatography-mass spectrometry (SPME-GC-MS). The presence and persistence of VOCs were monitored in 30 minute intervals, over a 4 hour period, in a triplicate time study. As predicted, spray paint solvents are lost to the environment readily, whereas few VOCs remained present in the headspace in low concentrations beyond 4 hours. The VOCs that were observed to have the highest persistence in the headspace were aromatic compounds and those with longer hydrocarbon chains. We present this study in a forensic science context and suggest that the interpretation of the results may be useful for forensic applications in establishing a time since deposition of a spray-painted surface.