Merck
  • Gold Nanoparticles and Graphene Oxide Flakes Synergistic Partaking in Cytosolic Bactericidal Augmentation: Role of ROS and NOX2 Activity.
Gold Nanoparticles and Graphene Oxide Flakes Synergistic Partaking in Cytosolic Bactericidal Augmentation: Role of ROS and NOX2 Activity.
Microorganisms (2021-01-21)
Osamah Al Rugaie, Majid Jabir, Rua Kadhim, Esraa Karsh, Ghassan M Sulaiman, Salman A A Mohammed, Riaz A Khan, Hamdoon A Mohammed
ABSTRAKT
Gold nanoparticles (GNPs) and graphene oxide flakes (GOFs) exerted significantly (p < 0.0001) supportive roles on the phagocytosis bioactivity of the immune cells of phagocytic nature against the Gram-positive and Gram-negative human pathogenic bacteria Staphylococcus aureus and Escherichia coli. Under experimental conditions, upon bacterial exposure, the combined GNPs and GOFs induced significant clearance of bacteria through phagosome maturation (p < 0.0001) from time-points of 6 to 30 min and production of reactive oxygen species (ROS, p < 0.0001) through the NADPH oxidase 2 (NOX2, p < 0.0001)-based feedback mechanism. The effects of the combined presence of GNPs and GOFs on phagocytosis (p < 0.0001) suggested a synergistic action underway, also achieved through elevated signal transduction activity in the bone-marrow-derived macrophages (BMDM, p < 0.0001). The current study demonstrated that GNPs' and GOFs' bactericidal assisting potentials could be considered an effective and alternative strategy for treating infections from both positive and negative bacterial strains.