Merck
  • 'Urease immobilized single-kit' for sensing of thiourea-glucose pair employing fluorescence 'Turn off - Turn on' and as an efficient sorbent for selective sample cleanup of thiourea.
'Urease immobilized single-kit' for sensing of thiourea-glucose pair employing fluorescence 'Turn off - Turn on' and as an efficient sorbent for selective sample cleanup of thiourea.
Analytica chimica acta (2020-11-30)
Sneha Mondal, Sanchari Saha, Shailja Mishra, Rimi Sarkar, Dipika Roy, Bhabatosh Mandal
ABSTRAKT
The tenfold lowering in binding energy for TU-Tyrosine in immobilized urease (Kb: 4.7 × 103) with respect to the native enzyme (Kb: 6.5 × 104) begets easy desorption of thiourea (TU) by glucose (GL) with an eventual formation of a more strong TU- GL adduct; that rejuvenates the kit-material ready for the subsequent cycle(s). The sorption-desorption heeds fluorescence turn-off and turn-on in DCM for selective sensing of TU- GL pair at their respective linear range of concentration 2.5-26.1 ppm and 2.36-11.57 ppm. The process was found to be static (KSV ≥ 2.25 × 103 L mol-1), exothermic (ΔH: -0.08 kJ mol-1), spontaneous (ΔG: -21.1 kJ mol-1) and marginally entropy gaining (ΔS: 0.07 kJ mol-1 K-1). The 'bulk material' (200 ± 20 μm) brilliantly preconcentrates TU with an enrichment factor of 106.2 after its selective extraction at near-neutral pH from a large volume sample (800 mL) of low concentration (30 ppm). A very dilute solution (0.05 mmol L-1) of GL at minimum volume (6 mL) acts as a stripping agent and provides a longer life (200 cycles with good extraction efficiency) to the material. The method was found to be efficient in the analysis of fruit juice as a real sample.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)