Merck
Platelet Apoptotic Response May Be Associated With the Capacity of Aspirin to Inhibit Platelets.
Journal of cardiovascular pharmacology (2020-11-11)
José J Zamorano-León, Martin Gascón, Carlos H Martínez, Gala Freixer, Redy Guerra, Khaoula Zekri-Nechar, Esther Bernardo, Mariano de la Serna-Soto, Antonio Segura, Manel Giner, Miguel A García-Fernández, Carlos Macaya, Antonio J López-Farré
ABSTRAKT
An inadequate platelet response to aspirin (ASA) has been identified in some patients under chronic ASA treatment. The aim of this study was to analyze if ASA-sensitive and ASA-resistant platelets have differences in their apoptotic capability. Clinically stable ischemic coronary patients who had been taking ASA (100 mg/d) for at least 9 months before inclusion were divided into ASA-resistant (n = 11) and ASA-sensitive (n = 13) groups as defined by the PFA-100 test. Platelets from ASA-sensitive patients showed higher expression of the proapoptotic proteins Bak and Bax than those from ASA-resistant patients, although only Bak protein remained different when the results were adjusted by age. In resting platelets, neither caspase-3 activity nor cytosolic cytochrome C levels were different between both experimental groups. Stimulation of platelets with calcium ionophore (10 nmol/L, A23187) increased caspase-3 activity (1.91-fold higher; P < 0.05) and cytosolic cytochrome C levels (1.84-fold higher; P < 0.05) to a higher degree in ASA-sensitive than in ASA-resistant platelets. In conclusion, ASA-sensitive platelets seem to be better prepared to undergo apoptosis during robust platelet activation.