Merck
Phthalates in House and Dormitory Dust: Occurrence, Human Exposure and Risk Assessment.
Bulletin of environmental contamination and toxicology (2020-11-29)
Shen Xu, Chao Li
ABSTRAKT
Phthalates are one of ubiquitous contaminants in the indoor environment. In this study, we analyzed concentrations and profiles of 9 phthalates in dust samples collected from houses and university dormitories in Nanjing, China. The total concentrations of phthalates in house and dormitory dust ranged from 111.4 to 3599.1 µg/g and 86.1 to 1262.3 µg/g, respectively. Phthalates in house was significantly higher than that in dormitory dust (p < 0.01). Di(2-ethylhexyl) phthalate (DEHP), dibutyl phthalate (DBP) and di-isobutyl phthalate (DiBP) were the three predominant compounds and accounted for more than 98% of total phthalates in the two microenvironments. The estimated daily intake (EDI) of phthalates, carcinogenic risk (CR) of DEHP, and hazard index (HI) values of DEHP, DBP and DiBP were estimated. Except for adults, the CR of DEHP for four subgroups (infants, toddlers, children, and teenagers) had exceeded the limitation, implying that they are at the risk of exposure to DEHP through dust ingestion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dess-Martin periodinane, 97%
Supelco
Dihexyl phthalate, analytical standard