Merck
In silico study, synthesis, and evaluation of the antimalarial activity of hybrid dimethoxy pyrazole 1,3,5-triazine derivatives.
Journal of biochemical and molecular toxicology (2020-12-18)
Pinku Gogoi, Anshul Shakya, Surajit K Ghosh, Neelutpal Gogoi, Prashant Gahtori, Nardev Singh, Dibya R Bhattacharyya, Udaya P Singh, Hans R Bhat
ABSTRAKT
Malaria continues to become a major global health problem, particularly in Sub-Saharan Africa, Asia, and Latin America. The widespread emergence of resistance to first-line drugs has further bolstered an urgent need for a new and cost-effective antimalarial(s). Thus, the present study enumerates the synthesis of novel hybrid dimethoxy pyrazole 1,3,5-triazine derivatives 7(a-j) and their in silico results short-listed three compounds with good binding energies and dock scores. Docking analysis shows that hydrogen-bonding predominates and typically involves key residues, such as Asp54, Tyr170, Ile164, and Arg122. The in vitro antimalarial evaluation of three top-ranked compounds (7e, 7g, and 7h) showed half-maximal inhibitory concentration values range from 53.85 to 100 μg/ml against chloroquine-sensitive strain 3D7 of Plasmodium falciparum. Compound 7e may be utilized as a lead for further optimization work in drug discovery due to good antimalarial activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propylene sulfite, 99%