Merck
A novel alkaloid glycoside isolated from Atalantia buxifolia.
Natural product research (2019-05-30)
Hui-Xian Liang, Jing-Jing Sun, Zhi-Bin Shen, Bang-Wei Yu, Hong-Hua Cui, Yong-Qin Yin
ABSTRAKT
From the dried root and rhizomes of Atalantia buxifolia afford 11 compounds, one new compound 1 and ten known compounds 2-11 were isolated and identified. The novel compound 1 was alkaloid glycoside. Its mother nuclear was the acridone, which was relatively rare. The structure of compound 1 was established identified by spectrum and elucidated as β-D-Glu-4,5-dimethoxy-1,6-dihydroxy-10-methyl-acridone. The compound 1 enriched the compound library and laid the material foundation for the subsequent study of pharmacological activities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propylene sulfite, 99%