Merck
  • Purification of Gekko Small Peptide Fraction and Its Effect of Inducing Apoptosis of EC 9706 Esophageal Cancer Cells by Inhibiting PI3K/Akt/GLUT1 Signaling Pathway.
Purification of Gekko Small Peptide Fraction and Its Effect of Inducing Apoptosis of EC 9706 Esophageal Cancer Cells by Inhibiting PI3K/Akt/GLUT1 Signaling Pathway.
Chemistry & biodiversity (2021-02-04)
Xingzhi Lv, Ruifang Li, Zhongjie Li, Jiangang Wang
ABSTRAKT
This study aimed to isolate and purify a cytotoxic extraction from Gekko japonicus, identify its components and determine its cytotoxic activity in vitro. We isolated and identified the most potent cytotoxic Gekko small peptide LH-20-15. The identification and analysis of peptide sequences of LH-20-15 were performed by de novo peptide sequencing, and two new peptides were found. LH-20-15 significantly inhibited the proliferation of human esophageal squamous carcinoma EC 9706 cells in a dose-dependent manner. Furthermore, LH-20-15 induced apoptosis in esophageal cancer cells by activating the mitochondrial apoptotic pathway. Further research showed that LH-20-15 inhibited the PI3 K/Akt/GLUT1 signaling pathway. In conclusion, LH-20-15 from Gekko japonicus is a peptide mixture and may inhibit EC 9706 cell proliferation and induce apoptosis by activating the mitochondrial apoptotic pathway. It also regulates glucose metabolism by targeting the PI3 K/Akt/GLUT1 signaling pathway. These small peptides could be new sources of natural cytotoxic ingredients against esophageal cancer with potential drug values.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
JC-1, powder or solid (Crystals)