Merck
  • Nucleoside transport inhibitors: structure-activity relationships for pyrimido[5,4-d]pyrimidine derivatives that potentiate pemetrexed cytotoxicity in the presence of α1-acid glycoprotein.
Nucleoside transport inhibitors: structure-activity relationships for pyrimido[5,4-d]pyrimidine derivatives that potentiate pemetrexed cytotoxicity in the presence of α1-acid glycoprotein.
Journal of medicinal chemistry (2011-03-04)
Kappusamy Saravanan, Hannah C Barlow, Marion Barton, A Hilary Calvert, Bernard T Golding, David R Newell, Julian S Northen, Nicola J Curtin, Huw D Thomas, Roger J Griffin
ABSTRAKT
Membrane transport of nucleosides or nucleobases is mediated by transporters including the equilibrative nucleoside transporters (ENTs), and resistance to antitumor antimetabolite drugs may arise via salvage of exogenous purine or pyrimidine nucleosides or nucleobases by ENT transporters. The therapeutic utility of dipyridamole (3), a potent ENT inhibitor, is compromised by binding to the serum protein α(1)-acid glycoprotein (AGP). Derivatives and prodrugs of the ENT inhibitor 4,8-bis[(3,4-dimethoxybenzyl)amino]-2,6-bis[(2-hydroxypropyl)amino]pyrimido[5,4-d]pyrimidine (6, NU3108) are described, with improved in vivo pharmacokinetic properties and reduced AGP binding relative to dipyridamole. The mono- and diglycine carbamate derivatives were at least as potent as 6 and showed no reduction in potency by AGP. In a [(3)H]thymidine incorporation assay, employing COR-L23 cells, the diastereoisomers of 6 (IC(50) = 26 nM) exhibited activity comparable with 3 (IC(50) = 15 nM). The monophenyl carbamate and mono-4-methoxyphenyl carbamate exhibited the best ENT-inhibitory activity in the COR-L23 assay (IC(50) = 8 and 4 nM, respectively). All of the new prodrugs were also highly effective at reversing thymidine/hypoxanthine rescue from pemetrexed cytotoxicity in the COR-L23 cell line.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dipyridamole, ≥98% (HPLC)
COR-L23, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 92031919, human lung, Epithelial