Merck
Inhibitory CD161 receptor identified in glioma-infiltrating T cells by single-cell analysis.
Cell (2021-02-17)
Nathan D Mathewson, Orr Ashenberg, Itay Tirosh, Simon Gritsch, Elizabeth M Perez, Sascha Marx, Livnat Jerby-Arnon, Rony Chanoch-Myers, Toshiro Hara, Alyssa R Richman, Yoshinaga Ito, Jason Pyrdol, Mirco Friedrich, Kathrin Schumann, Michael J Poitras, Prafulla C Gokhale, L Nicolas Gonzalez Castro, Marni E Shore, Christine M Hebert, Brian Shaw, Heather L Cahill, Matthew Drummond, Wubing Zhang, Olamide Olawoyin, Hiroaki Wakimoto, Orit Rozenblatt-Rosen, Priscilla K Brastianos, X Shirley Liu, Pamela S Jones, Daniel P Cahill, Matthew P Frosch, David N Louis, Gordon J Freeman, Keith L Ligon, Alexander Marson, E Antonio Chiocca, David A Reardon, Aviv Regev, Mario L Suvà, Kai W Wucherpfennig
ABSTRAKT
T cells are critical effectors of cancer immunotherapies, but little is known about their gene expression programs in diffuse gliomas. Here, we leverage single-cell RNA sequencing (RNA-seq) to chart the gene expression and clonal landscape of tumor-infiltrating T cells across 31 patients with isocitrate dehydrogenase (IDH) wild-type glioblastoma and IDH mutant glioma. We identify potential effectors of anti-tumor immunity in subsets of T cells that co-express cytotoxic programs and several natural killer (NK) cell genes. Analysis of clonally expanded tumor-infiltrating T cells further identifies the NK gene KLRB1 (encoding CD161) as a candidate inhibitory receptor. Accordingly, genetic inactivation of KLRB1 or antibody-mediated CD161 blockade enhances T cell-mediated killing of glioma cells in vitro and their anti-tumor function in vivo. KLRB1 and its associated transcriptional program are also expressed by substantial T cell populations in other human cancers. Our work provides an atlas of T cells in gliomas and highlights CD161 and other NK cell receptors as immunotherapy targets.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.