Merck
Involvement of autophagy in realgar quantum dots (RQDs) inhibition of human endometrial cancer JEC cells.
PeerJ (2020-11-06)
Zhengyun Liu, Ke Xu, Yan Xu, Wanling Zhang, Nian Jiang, Shengyu Wang, Guo Luo, Jie Liu, Jinzhu Wu, Huan Wang
ABSTRAKT
Realgar (As4S4) has been used in traditional Chinese medicines for treatment of malignancies. The poor solubility of As4S4 hampered its clinical applications. Realgar quantum dots (RQDs) were developed to overcome these problems. Previous studies revealed that the RQDs were effective against endometrial cancer JEC cells and hepatocarcinoma HepG2 cells via inducing apoptosis.Apoptosis and autophagy are important programmed cell death pathways leading to anticancer effects. This study further examined effects of RQDs on autophagy, focusing on the formation of the autophagosome in JEC cells. CCK8 assay was used to examine cell proliferation. Flow cytometry was used to analyze cell cycle. Transmission electron microscopy (TEM) was used to examine the autophagy, cells were transfected with pEGFP-C3-MAP1LC3B plasmid to examine effects of RQDs on autophagosome via confocal microscope. Autophagy-related proteins were examined by Western blot. RQDs exhibited cytotoxicity in JEC cells in a concentration- and time- dependent manner. RQDs induced G2 and S phase arrest in JEC cells. RQDs significantly induced autophagy, with the double-membrane and autophagosome-like structures by TEM. The diffused distribution of pEGFP-C3-MAP1LC3B green fluorescence were become the punctuate pattern fluorescence after treatment with RQDs in cells transfected with pEGFP-C3-MAP1LC3B plasmid RQDs increased the expression of autophagyregulatory proteins LC3 I/II, Beclin-1, p62 and Atg12 in a concentration-dependent manner, similar to autophagy induced by serum starvation, except for p62, as induction of p62 is a characteristic of arsenic compounds. Taken together, the present study clearly demonstrated that RQDs can induce autophagy in JEC cells as one of mechanisms of anticancer effects, and indicated that RQDs may be developed as an autophagy inducer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D.E.R. 332, used as embedding medium