Merck
Effect of RIP Overexpression on Abiotic Stress Tolerance and Development of Rice.
International journal of molecular sciences (2021-02-05)
Pieter Wytynck, Jeroen Lambin, Simin Chen, Sinem Demirel Asci, Isabel Verbeke, Jeroen De Zaeytijd, Kondeti Subramanyam, Els J M Van Damme
ABSTRAKT
Ribosome-inactivating proteins (RIPs) are a class of cytotoxic enzymes that can inhibit protein translation by depurinating rRNA. Most plant RIPs are synthesized with a leader sequence that sequesters the proteins to a cell compartment away from the host ribosomes. However, several rice RIPs lack these signal peptides suggesting they reside in the cytosol in close proximity to the plant ribosomes. This paper aims to elucidate the physiological function of two nucleocytoplasmic RIPs from rice, in particular, the type 1 RIP referred to as OsRIP1 and a presumed type 3 RIP called nuRIP. Transgenic rice lines overexpressing these RIPs were constructed and studied for developmental effects resulting from this overexpression under greenhouse conditions. In addition, the performance of transgenic seedlings in response to drought, salt, abscisic acid and methyl jasmonate treatment was investigated. Results suggest that both RIPs can affect methyl jasmonate mediated stress responses.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)