Merck
  • Sex-dependent effects of social status on the regulation of arginine-vasopressin (AVP) V1a, oxytocin (OT), and serotonin (5-HT) 1A receptor binding and aggression in Syrian hamsters (Mesocricetus auratus).
Sex-dependent effects of social status on the regulation of arginine-vasopressin (AVP) V1a, oxytocin (OT), and serotonin (5-HT) 1A receptor binding and aggression in Syrian hamsters (Mesocricetus auratus).
Hormones and behavior (2020-11-06)
Z A Grieb, A P Ross, K E McCann, S Lee, M Welch, M G Gomez, A Norvelle, V Michopoulos, K L Huhman, H E Albers
ABSTRAKT
Dominance status in hamsters is driven by interactions between arginine-vasopressin V1a, oxytocin (OT), and serotonin 1A (5-HT1A) receptors. Activation of V1a and OT receptors in the anterior hypothalamus (AH) increases aggression in males, while decreasing aggression in females. In contrast, activation of 5-HT1A receptors in the AH decreases aggression in males and increases aggression in females. The mechanism underlying these differences is not known. The purpose of this study was to determine if dominance status and sex interact to regulate V1a, OT, and 5-HT1A receptor binding. Same-sex hamsters (N = 47) were paired 12 times across six days in five min sessions. Brains from paired and unpaired (non-social control) hamsters were collected immediately after the last interaction and processed for receptor binding using autoradiography. Differences in V1a, OT, and 5-HT1A receptor binding densities were observed in several brain regions as a function of social status and sex. For example, in the AH, there was an interaction between sex and social status, such that V1a binding in subordinate males was lower than in subordinate females and V1a receptor density in dominant males was higher than in dominant females. There was also an interaction in 5-HT1A receptor binding, such that social pairing increased 5-HT1A binding in the AH of males but decreased 5-HT1A binding in females compared with unpaired controls. These results indicate that dominance status and sex play important roles in shaping the binding profiles of key receptor subtypes across the neural circuitry that regulates social behavior.