Merck
Evaluation of dihydropyrimidin-(2H)-one analogues and rhodanine derivatives as tyrosinase inhibitors.
Bioorganic & medicinal chemistry letters (2011-03-18)
Jinbing Liu, Fengyan Wu, Lingjuan Chen, Jianming Hu, Liangzhong Zhao, Changhong Chen, Liwang Peng
ABSTRAKT
A series of dihydropyrimidin-(2H)-one analogues and rhodanine derivatives were synthesized and their inhibitory effects on the diphenolase activity of mushroom tyrosinase were evaluated. The results showed that some of the synthesized compounds exhibited significant inhibitory activities. Especially, compound 15 bearing a hydroxyethoxyl group at position-4 of phenyl ring exhibited most potent tyrosinase inhibitory activity with IC(50) value of 0.56 mM. The inhibition mechanism analysis of compound 15 demonstrated that the inhibitory effect of the compound on the tyrosinase was irreversible. These results suggested that such compounds might be served as lead compounds for further designing new potential tyrosinase inhibitors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%