Merck
N-glycomic Analysis of Z(IgA1) Partitioned Serum and Salivary Immunoglobulin A by Capillary Electrophoresis.
Current molecular medicine (2020-04-14)
Brigitta Meszaros, Zsuzsanna Kovacs, Eniko Gebri, Hajnalka Jankovics, Ferenc Vonderviszt, Attila Kiss, Adam Simon, Sarolta Botka, Tibor Hortobagyi, Andras Guttman
ABSTRAKT
Application of capillary electrophoresis with laser induced fluorescence detection (CE-LIF) to identify the N-glycosylation structures of serum and saliva IgA from healthy controls and patients with malignant hematological diseases having cytostatic treatment induced mild oral mucosal lesions. Altered N-glycosylation of body fluid glycoproteins can be an effective indicator of most inflammatory processes. Immunoglobulin A (IgA) is the second highest abundant immunoglobulin and has a major role in the immune-defense against potential pathogen attacks. While IgA is abundant in serum, secretory immunoglobulin A (sIgA) is one of the most prevalent proteins in mucosal surfaces, such as in saliva. Our aim was to investigate the changes of IgA glycosylation in serum and saliva as a response to an administered cytostatic treatment in patients with malignant hematological disorders. Capillary electrophoresis with laser induced fluorescent detection (CE-LIF) was used to analyze the N-glycosylation profiles of Z(IgA1) partitioned immunoglobulin A in pooled serum and saliva of 10 control subjects and 8 patients with malignant hematological diseases having cytostatic treatment induced mild oral mucosal lesions. Eight of 31 and four of 38 N-glycans in serum and saliva, respectively, showed significant (p<0.05) differences upon comparison to the control group. Thirteen glycans were present in the saliva but not in the serum, on the other hand, six structures were found in the serum samples not present in the saliva. The developed Z(IgA1) partitioning and the high resolution CE-LIF based glyocoanalytical methods provided an efficient and sensitive workflow to detect and monitor IgA glycosylation alterations in serum and saliva with the scope for widespread molecular medicinal use.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology