Merck
The Effects of Premature Tooth Extraction and Damage on Replacement Timing in the Green Iguana.
Integrative and comparative biology (2020-09-26)
Kirstin S Brink, Ping Wu, Cheng-Ming Chuong, Joy M Richman
ABSTRAKT
Reptiles with continuous tooth replacement, or polyphyodonty, replace their teeth in predictable, well-timed waves in alternating tooth positions around the mouth. This process is thought to occur irrespective of tooth wear or breakage. In this study, we aimed to determine if damage to teeth and premature tooth extraction affects tooth replacement timing long-term in juvenile green iguanas (Iguana iguana). First, we examined normal tooth development histologically using a BrdU pulse-chase analysis to detect label-retaining cells in replacement teeth and dental tissues. Next, we performed tooth extraction experiments for characterization of dental tissues after functional tooth (FT) extraction, including proliferation and β-Catenin expression, for up to 12 weeks. We then compared these results to a newly analyzed historical dataset of X-rays collected up to 7 months after FT damage and extraction in the green iguana. Results show that proliferation in the dental and successional lamina (SL) does not change after extraction of the FT, and proliferation occurs in the SL only when a tooth differentiates. Damage to an FT crown does not affect the timing of the tooth replacement cycle, however, complete extraction shifts the replacement cycle ahead by 4 weeks by removing the need for resorption of the FT. These results suggest that traumatic FT loss affects the timing of the replacement cycle at that one position, which may have implications for tooth replacement patterning around the entire mouth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid