Merck
  • Polyacrylonitrile-based carbon nanofibers as a matrix for laser desorption/ionization time-of-flight mass spectrometric analysis of small molecules under both positive and negative ionization modes.
Polyacrylonitrile-based carbon nanofibers as a matrix for laser desorption/ionization time-of-flight mass spectrometric analysis of small molecules under both positive and negative ionization modes.
Analytical and bioanalytical chemistry (2021-01-07)
Ari Chae, Gwanwon Lee, Dong-Yeun Koh, Cheol-Min Yang, Sungho Lee, Young-Kwan Kim
ABSTRAKT
Carbon fiber (CNF), prepared by carbonization of electrospun polyacrylonitrile (PAN) fibers, is systematically investigated as a mediator to replace conventional organic matrices for matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-MS). CNF exhibits a high salt tolerance, sensitivity, and resolution for organic matrix-free laser desorption/ionization time-of-flight mass spectrometry (LDI-MS) analysis of various analytes under both positive and negative ionization modes. Especially, saccharides, a neutral molecule having low negative ionization efficiency, are successfully detected with CNF. Taken together, this study clearly demonstrates CNF is a promising material to develop an efficient and universal platform for LDI-MS analysis regardless of preferential ionization modes of analytes. Graphical abstract.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Yttrium aluminum oxide, nanopowder, <150 nm particle size (TEM), 99% trace metals basis