Merck
DIPPER, a spatiotemporal proteomics atlas of human intervertebral discs for exploring ageing and degeneration dynamics.
eLife (2021-01-01)
Vivian Tam, Peikai Chen, Anita Yee, Nestor Solis, Theo Klein, Mateusz Kudelko, Rakesh Sharma, Wilson Cw Chan, Christopher M Overall, Lisbet Haglund, Pak C Sham, Kathryn Song Eng Cheah, Danny Chan
ABSTRAKT
The spatiotemporal proteome of the intervertebral disc (IVD) underpins its integrity and function. We present DIPPER, a deep and comprehensive IVD proteomic resource comprising 94 genome-wide profiles from 17 individuals. To begin with, protein modules defining key directional trends spanning the lateral and anteroposterior axes were derived from high-resolution spatial proteomes of intact young cadaveric lumbar IVDs. They revealed novel region-specific profiles of regulatory activities and displayed potential paths of deconstruction in the level- and location-matched aged cadaveric discs. Machine learning methods predicted a 'hydration matrisome' that connects extracellular matrix with MRI intensity. Importantly, the static proteome used as point-references can be integrated with dynamic proteome (SILAC/degradome) and transcriptome data from multiple clinical samples, enhancing robustness and clinical relevance. The data, findings, and methodology, available on a web interface (http://www.sbms.hku.hk/dclab/DIPPER/), will be valuable references in the field of IVD biology and proteomic analytics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lys-Lys-Lys-Lys, ≥95% (TLC)