Merck
  • Modulating effect of hesperetin on the molecular expression pattern of apoptotic and cell proliferative markers in 7,12-dimethylbenz(a)anthracene-induced oral carcinogenesis.
Modulating effect of hesperetin on the molecular expression pattern of apoptotic and cell proliferative markers in 7,12-dimethylbenz(a)anthracene-induced oral carcinogenesis.
Archives of physiology and biochemistry (2019-01-22)
Sukumar Babukumar, Veerasamy Vinothkumar, Duraisamy Ramachandhiran
ABSTRAKT
Oral squamous cell carcinoma (OSCC) is widespread malignant neoplasm and refractory cancers in worldwide. Here, we studied the chemopreventive potential of hesperetin on 7,12-dimethylbenz(a)anthracene (DMBA)-induced hamster buccal pouch carcinogenesis (HBPCs). Oral tumours were developed in the buccal pouches of male golden Syrian hamsters by topical application of 0.5% DMBA three times a week for 10 weeks. This causes sequentially hyperplasia, dysplasia and well differentiates squamous cell carcinoma (SCC) with up-regulation of molecular markers like mutant-p53, Caspase-3 and caspase-9 and cyclin-D1. Histology, immunohistochemistry (IHC), real time PCR (qRT-PCR) and western blot analysis of hesperetin treated animals shows a reversal in the above expression pattern to near normal in buccal mucosal tissue. Therefore, hesperetin exhibits the potential protective effect against DMBA-induced oral cancer through apoptotic and anti-proliferative properties. However, a long-term observation would be needed to confirm the possibility of malignant change of the resulted dysplastic lesions upon hesperetin pretreatment.