Merck
Rheological performance of film-forming solutions made from plasma-modified starches with different amylose/amylopectin content.
Carbohydrate polymers (2021-01-14)
Pablo Hernandez-Perez, Pamela C Flores-Silva, Gonzalo Velazquez, Eduardo Morales-Sanchez, Oliverio Rodríguez-Fernández, Ernesto Hernández-Hernández, Guadalupe Mendez-Montealvo, Israel Sifuentes-Nieves
ABSTRAKT
Normal and high amylose corn starches were modified using HMDSO plasma at different time treatments. Changes in functional properties of starch granule, film-forming solutions (FFS) and films were investigated. SEM analysis revealed HMDSO coating deposition on the granule surface, which limited the amylopectin leach out from the granules to the continuous matrix, affecting the rheological properties of the FFS. The amylopectin restriction resulted in a low reinforcement of the network decreasing the viscosity as indicated by n and k values. Also, a gel-like behavior (G' > G″) was observed when the amylose and time treatment increased, suggesting that the matrix becomes less elastic with softer entanglement. This behavior was confirmed by creep test and Burger model parameters. The plasma treatments allowed obtaining FFS with low viscosity, suitable for developing soft and hydrophobic films with low flexibility, as indicated by the decrease of the maximum stress, Hencky strain and permeance values.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hexamethyldisiloxane, ≥98%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)