Merck
  • Development of the spatial contrast sensitivity function (CSF) during childhood: Analysis of previous findings and new psychophysical data.
Development of the spatial contrast sensitivity function (CSF) during childhood: Analysis of previous findings and new psychophysical data.
Journal of vision (2020-12-05)
Tessa M Dekker, Mahtab Farahbakhsh, Janette Atkinson, Oliver J Braddick, Pete R Jones
ABSTRAKT
Although the contrast sensitivity function (CSF) changes markedly during infancy, there is no consensus regarding whether, how, and why it continues to develop in later childhood. Here, we analyzed previously published data (N = 1928 CSFs), and present new psychophysical findings from 98 children (4.7-14.8 years) and 50 adults (18.1-29.7 years), in order to answer the following questions: (1) Does the CSF change during childhood? (2) How large is the developmental effect size? (3) Are any changes uniform across the CSF, or frequency-specific? and (4) Can some or all of the changes be explained by "non-visual" (i.e. procedural/cognitive) factors, such as boredom or inattentiveness? The new data were collected using a four-alternative forced-choice (4AFC) Gabor-detection task, with two different psychophysical procedures (Weighted Staircase; QUEST+), and suprathreshold (false-negative) catch trials to quantify lapse rates. It is shown that from ages 4 to 18 years, the CSF improves (at an exponentially decaying rate) by approximately 0.3 log10 units (a doubling of contrast sensitivity [CS]), with 90% of this change complete by 12 years of age. The size of the effect was small relative to individual variability, with age alone explaining less than one sixth of variability (16%), and most children performing as well as some adults (i.e. falling within the 90% population limits for adults). Development was frequency-specific, with changes occurring primarily around or below the CSF peak (≤ 4 cpd). At least half - and potentially all - of the changes observed could be explained by non-visual factors (e.g. lapses in concentration), although possible biological mechanisms are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Zinc isopropylsulfinate, 95%