Merck
The choice of biopolymer is crucial to trigger angiogenesis with vascular endothelial growth factor releasing coatings.
Journal of materials science. Materials in medicine (2020-10-28)
Christiane Claaßen, Miriam Dannecker, Jana Grübel, Maria-Elli Kotzampasi, Günter E M Tovar, Boris V Stanzel, Kirsten Borchers
ABSTRAKT
Bio-based coatings and release systems for pro-angiogenic growth factors are of interest to overcome insufficient vascularization and bio-integration of implants. This study compares different biopolymer-based coatings on polyethylene terephthalate (PET) membranes in terms of coating homogeneity and stability, coating thickness in the swollen state, endothelial cell adhesion, vascular endothelial growth factor (VEGF) release and pro-angiogenic properties. Coatings consisted of carbodiimide cross-linked gelatin type A (GelA), type B (GelB) or albumin (Alb), and heparin (Hep), or they consisted of radically cross-linked gelatin methacryloyl-acetyl (GM5A5) and heparin methacrylate (HepM5). We prepared films with thicknesses of 8-10 µm and found that all coatings were homogeneous after washing. All gelatin-based coatings enhanced the adhesion of primary human endothelial cells compared to the uncoated membrane. The VEGF release was tunable with the loading concentration and dependent on the isoelectric points and hydrophilicities of the biopolymers used for coating: GelA-Hep showed the highest releases, while releases were indistinguishable for GelB-Hep and Alb-Hep, and lowest for GM5A5-HepM5. Interestingly, not only the amount of VEGF released from the coatings determined whether angiogenesis was induced, but a combination of VEGF release, metabolic activity and adhesion of endothelial cells. VEGF releasing GelA-Hep and GelB-Hep coatings induced angiogenesis in a chorioallantoic membrane assay, so that these coatings should be considered for further in vivo testing.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥98%