Merck
  • Substituted phenyl 4-(2-oxoimidazolidin-1-yl)benzenesulfonamides as antimitotics. Antiproliferative, antiangiogenic and antitumoral activity, and quantitative structure-activity relationships.
Substituted phenyl 4-(2-oxoimidazolidin-1-yl)benzenesulfonamides as antimitotics. Antiproliferative, antiangiogenic and antitumoral activity, and quantitative structure-activity relationships.
European journal of medicinal chemistry (2011-09-17)
Sébastien Fortin, Lianhu Wei, Emmanuel Moreau, Jacques Lacroix, Marie-France Côté, Eric Petitclerc, Lakshmi P Kotra, René C Gaudreault
ABSTRAKT
The importance of the bridge linking the two phenyl moieties of substituted phenyl 4-(2-oxoimidazolidin-1-yl)benzenesulfonates (PIB-SOs) was assessed using a sulfonamide group, which is a bioisostere of sulfonate and ethenyl groups. Forty one phenyl 4-(2-oxoimidazolidin-1-yl)benzenesulfonamide (PIB-SA) derivatives were prepared and biologically evaluated. PIB-SAs exhibit antiproliferative activities at the nanomolar level against sixteen cancer cell lines, block the cell cycle progression in G(2)/M phase, leading to cytoskeleton disruption and anoikis. These results were subjected to CoMFA and CoMSIA analyses to establish quantitative structure-activity relationships. These results evidence that the sulfonate and sulfonamide moieties are reciprocal bioisosteres and that phenylimidazolidin-2-one could mimic the trimethoxyphenyl moiety found in the structure of numerous potent antimicrotubule agents. Finally, compounds 16 and 17 exhibited potent antitumor and antiangiogenic activities on HT-1080 fibrosarcoma cells grafted onto chick chorioallantoic membrane similar to CA-4 without significant toxicity for the chick embryos, making this class of compounds a promising class of anticancer agents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Colchicine, suitable for plant cell culture, BioReagent, ≥95% (HPLC)