Merck
RIPK2 is an unfavorable prognosis marker and a potential therapeutic target in human kidney renal clear cell carcinoma.
Aging (2021-04-02)
Diangeng Li, Liangyou Tang, Bo Liu, Shibo Xu, Meiling Jin, Wu Bo
ABSTRAKT
Receptor Interacting Serine/Threonine Kinase 2 (RIPK2) is located on chromosome 8q21 and encodes a protein containing a C-terminal caspase activation and recruitment domain (CARD), which is a component of signaling complexes in both the innate and adaptive immune pathways. To estimate the value of RIPK2 in evaluating the prognosis and guiding the targeted therapy for patients with kidney renal clear cell carcinoma (KIRC), we analyzed total 526 KIRC samples from The Cancer Genome Atlas (TCGA) database. Our result showed that RIPK2 was upregulated in KIRC tumor samples compared with normal samples. Cox regression was performed to calculate the hazard ratio of RIPK2 expression as an unfavorable prognosis feature for overall survival. Moreover, RIPK2 expression was positively correlated to the high-risk clinical stage, and metastasis features. The upregulation of RIPK2 was strongly correlated with various immune signaling pathway dysregulations as well as immune phenotypes changes in KIRC patient's cohort. In addition, inhibition of RIPK2 activity by either shRNA-mediated knockdown or inhibitor significantly reduced kidney cancer cell viability, trans-migration in vitro, and impaired tumor growth in vivo. In conclusion, elevated RIPK2 expression indicates a worse prognosis for KIRC patients and could serve as a potential prognostic biomarker and therapeutic target in kidney cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid