Merck
Identification and expression of a unique neonatal variant of the GABAA receptor α3 subunit.
Brain structure & function (2017-12-29)
Stephanie M Miller, Samuel Pelly, Viskasari P Kalanjati, Aven Lee, Paul B Colditz, S Tracey Bjorkman
ABSTRAKT
The GABAA receptor provides the majority of inhibitory neurotransmission in the adult central nervous system but in immature brain is responsible for much of the excitatory drive, a requirement for normal brain development. It is well established that GABAA receptor subunit expression changes across the course of brain development. In the present study, we have identified a splice variant of the GABAA receptor α3 subunit which appears unique to the developing brain, referred to here as the GABAA receptor α3 subunit neonatal variant (GABAA receptor α3N). RT-PCR and sequence analysis revealed splicing of exon 8 of the α3 subunit. Western blot analysis showed expression of GABAA receptor α3N in the cortex of several neonatal species and significantly reduced expression of this splice variant in the corresponding adult brains. Expression was evident in multiple brain regions and decreased across development in the pig. Fractionation revealed differential cellular localisation in the parietal cortex, hippocampus and thalamus of the full-length GABAA receptor α3 and GABAA receptor α3N. Immunoprecipitation showed direct interaction with the GABAA receptor subunits α1 and γ2 but not with gephyrin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-GABA A Receptor α1 chain Antibody, NT, clone BD24, clone BD24, Chemicon®, from mouse
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Przeciwciało przeciwko receptorowi γ2 GABA A, pętla cytoplazmatyczna, Chemicon®, from rabbit
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder