Merck
An expedient synthesis of honokiol and its analogues as potential neuropreventive agents.
Bioorganic & medicinal chemistry letters (2011-12-07)
Subhankar Tripathi, Ming-Huan Chan, Chinpiao Chen
ABSTRAKT
An efficient synthesis of honokiol with Suzuki-Miyaura cross coupling obtained an overall yield of 45%. The proposed approach successfully synthesized several structurally similar alkyl, alkenyl and alkynyl analogues, seven of which showed potential neuropreventive activity against MPP(+)-induced and CHP/TBHP oxidative stress induced neuroblastoma cell death.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O